ENA024295923 Enamine

5-amino-1-phenyl-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD00847752
  • Cas Number : 69923-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-178
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09887
Availability details for
Quantity
  • Availability
  • Add to Cart