ENA024295924 Enamine

1-phenyl-6-sulfanyl-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD06356487
  • Cas Number : 156718-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09888
Availability details for
Quantity
  • Availability
  • Add to Cart