ENA024295933 Enamine

2-(4-methoxybenzenesulfonamido)-3-methylbutanoic acid

  • MDL number MFCD04113951
  • Cas Number : 197965-89-2
  • Molecular Weight : 287.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-09897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09897
Availability details for
Quantity
  • Availability
  • Add to Cart