ENA024295948 Enamine

5-(2-chlorophenyl)-3-(2-methoxyethyl)-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD06356485
  • Cas Number : 790232-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09912
Availability details for
Quantity
  • Availability
  • Add to Cart