ENA024295951 Enamine

3-(3-methoxypropyl)-6-phenyl-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD06342814
  • Cas Number : 757221-93-5
  • Molecular Weight : 332.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09915
Availability details for
Quantity
  • Availability
  • Add to Cart