ENA024295958 Enamine

5-benzyl-4-(3-methylphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04087065
  • Cas Number : 76151-23-0
  • Molecular Weight : 281.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09922
Availability details for
Quantity
  • Availability
  • Add to Cart