ENA024295965 Enamine

6-amino-5-(dimethylamino)-1-methylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD06356461
  • Cas Number : 14094-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09929

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09929
Availability details for
Quantity
  • Availability
  • Add to Cart