ENA024295987 Enamine

4-cyclohexyl-5-[1-(dimethylamino)propyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06356501
  • Cas Number : 790232-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09951

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09951
Availability details for
Quantity
  • Availability
  • Add to Cart