ENA024295989 Enamine

2-[(4-bromophenyl)sulfanyl]pyridine-3-carboxylic acid

  • MDL number MFCD06340088
  • Cas Number : 955-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09953
Availability details for
Quantity
  • Availability
  • Add to Cart