ENA024295992 Enamine

ethyl 2-[(chloroacetyl)amino]-4-(2-furyl)thiophene-3-carboxylate

  • MDL number MFCD01917489
  • Cas Number : 356568-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09956

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-09956
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart