ENA024295995 Enamine

5-benzyl-4-(2-methoxyethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04141984
  • Cas Number : 663181-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09959
Availability details for
Quantity
  • Availability
  • Add to Cart