ENA024296006 Enamine

2-chloro-N-{[(2,3-dimethylcyclohexyl)amino]carbonyl}acetamide

  • MDL number MFCD06340181
  • Cas Number : 953807-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09970

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09970
Availability details for
Quantity
  • Availability
  • Add to Cart