ENA024296011 Enamine

methyl 5-{[(5-sulfanyl-1,3,4-thiadiazol-2-yl)sulfanyl]methyl}furan-2-carboxylate

  • MDL number MFCD06357811
  • Cas Number : 790270-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09975
Availability details for
Quantity
  • Availability
  • Add to Cart