ENA024296012 Enamine

3-chloro-5-methoxy-4-(propan-2-yloxy)benzoic acid

  • MDL number MFCD06197244
  • Cas Number : 749920-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09976
Availability details for
Quantity
  • Availability
  • Add to Cart