ENA024296021 Enamine

2-(hexanoylamino)-4,5-dimethoxybenzoic acid

  • MDL number MFCD06337030
  • Cas Number : 749920-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...180
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09985

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09985
Availability details for
Quantity
  • Availability
  • Add to Cart