ENA024296031 Enamine

4-(difluoromethoxy)-3-ethoxybenzoic acid

  • MDL number MFCD04967203
  • Cas Number : 162401-63-0
  • Molecular Weight : 232.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09995
Availability details for
Quantity
  • Availability
  • Add to Cart