ENA024296039 Enamine

5-{[3-(aminocarbonyl)thien-2-yl]amino}-5-oxopentanoic acid

  • MDL number MFCD03946373
  • Cas Number : 590376-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92...94
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10003

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10003
Availability details for
Quantity
  • Availability
  • Add to Cart