ENA024296040 Enamine

3-[(3-carbamoylthiophen-2-yl)carbamoyl]propanoic acid

  • MDL number MFCD03946372
  • Cas Number : 590376-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-172
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10004
Availability details for
Quantity
  • Availability
  • Add to Cart