ENA024296042 Enamine

2-chloro-N-(1-cyanocyclohexyl)-N-methylacetamide

  • MDL number MFCD05273565
  • Cas Number : 78734-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-79
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10006
Availability details for
Quantity
  • Availability
  • Add to Cart