ENA024296044 Enamine

2-chloro-N-{[(2-methoxy-5-methylphenyl)carbamoyl]methyl}acetamide

  • MDL number MFCD06337041
  • Cas Number : 749920-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-10008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10008
Availability details for
Quantity
  • Availability
  • Add to Cart