ENA024296049 Enamine

3-allyl-2-mercapto-4-oxo-3,4-dihydroquinazoline-7-carboxylic acid

  • MDL number MFCD02112591
  • Cas Number : 309969-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10013

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10013
Availability details for
Quantity
  • Availability
  • Add to Cart