ENA024296063 Enamine

3-(3-methylphenyl)-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD06357447
  • Cas Number : 819793-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10027
Availability details for
Quantity
  • Availability
  • Add to Cart