ENA024296075 Enamine

4-methyl-3-(morpholine-4-carbonyl)-5-phenylthiophen-2-amine

  • MDL number MFCD06337036
  • Cas Number : 749920-71-6
  • Molecular Weight : 302.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 170-171
Iupac Name 4-methyl-3-(4-morpholinylcarbonyl)-5-phenyl-2-thienylamine
Inchi Code 1S/C16H18N2O2S/c1-11-13(16(19)18-7-9-20-10-8-18)15(17)21-14(11)12-5-3-2-4-6-12/h2-6H,7-10,17H2,1H3
Inchi Key CEZSXQKFAJPXBR-UHFFFAOYSA-N

Description

Melting Point 170-171
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-10039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10039
Availability details for
Quantity
  • Availability
  • Add to Cart