ENA024296084 Enamine

2-(chloromethyl)-4H-3,1-benzoxazin-4-one

  • MDL number MFCD00100284
  • Cas Number : 98592-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91...93
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10048

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10048
Availability details for
Quantity
  • Availability
  • Add to Cart