ENA024296090 Enamine

1-mercapto-4-(3-methylbutyl)[1,2,4]triazolo[4,3-a]quinazolin-5(4H)-one

  • MDL number MFCD06337046
  • Cas Number : 750624-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10054

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10054
Availability details for
Quantity
  • Availability
  • Add to Cart