ENA024296099 Enamine

2-chloro-1-(4-ethoxyphenyl)propan-1-one

  • MDL number MFCD06357865
  • Cas Number : 790270-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10063
Availability details for
Quantity
  • Availability
  • Add to Cart