ENA024296105 Enamine

2-(2-amino-1,3-thiazol-4-yl)-1-phenylethan-1-one

  • MDL number MFCD02706789
  • Cas Number : 57626-32-1
  • Molecular Weight : 218.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225-226
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10069
Availability details for
Quantity
  • Availability
  • Add to Cart