ENA024296122 Enamine

3-[(2,3-dihydro-1,4-benzodioxin-6-yl)amino]propanenitrile

  • MDL number MFCD06340072
  • Cas Number : 757192-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-73
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10086
Availability details for
Quantity
  • Availability
  • Add to Cart