ENA024296135 Enamine

2-chloro-N-[1-(2-chlorophenyl)ethyl]acetamide

  • MDL number MFCD06357427
  • Cas Number : 40023-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10099
Availability details for
Quantity
  • Availability
  • Add to Cart