ENA024296138 Enamine

1-(4-fluorophenyl)-3-(4-methylphenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06357437
  • Cas Number : 956779-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202-203
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10102
Availability details for
Quantity
  • Availability
  • Add to Cart