ENA024296139 Enamine

1-[(4-ethyl-5-mercapto-4H-1,2,4-triazol-3-yl)methyl]azepan-2-one

  • MDL number MFCD06357820
  • Cas Number : 790270-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10103

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10103
Availability details for
Quantity
  • Availability
  • Add to Cart