ENA024296141 Enamine

2-(2-oxoazepan-1-yl)acetohydrazide

  • MDL number MFCD06357842
  • Cas Number : 790270-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10105
Availability details for
Quantity
  • Availability
  • Add to Cart