ENA024296144 Enamine

3-(3-methylphenyl)-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06357863
  • Cas Number : 956387-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-127
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10108
Availability details for
Quantity
  • Availability
  • Add to Cart