ENA024296146 Enamine

4-carbamoyl-2-[3-(trifluoromethyl)benzenesulfonamido]butanoic acid

  • MDL number MFCD05267034
  • Cas Number : 1008582-71-5
  • Molecular Weight : 354.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10110
Availability details for
Quantity
  • Availability
  • Add to Cart