ENA024296178 Enamine

({2-[(5-methylisoxazol-3-yl)amino]-2-oxoethyl}thio)acetic acid

  • MDL number MFCD00096559
  • Cas Number : 436091-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10142

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10142
Availability details for
Quantity
  • Availability
  • Add to Cart