ENA024296201 Enamine

1-(4-fluorophenyl)-3-(3-methylphenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06357819
  • Cas Number : 956181-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10165
Availability details for
Quantity
  • Availability
  • Add to Cart