ENA024296204 Enamine

{[5-(2-furyl)-1,3,4-oxadiazol-2-yl]thio}acetic acid

  • MDL number MFCD00450161
  • Cas Number : 33621-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152...155
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10168

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10168
Availability details for
Quantity
  • Availability
  • Add to Cart