ENA024296214 Enamine

5-amino-2-({[4-chloro-3-(trifluoromethyl)phenyl]sulfonyl}amino)-5-oxopentanoic acid

  • MDL number MFCD05269287
  • Cas Number : 1008984-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10178

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10178
Availability details for
Quantity
  • Availability
  • Add to Cart