ENA024296224 Enamine

4-(2-methylphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03900748
  • Cas Number : 36017-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-149
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10188
Availability details for
Quantity
  • Availability
  • Add to Cart