ENA024296233 Enamine

5-methyl-4-(2-methylphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02854377
  • Cas Number : 497079-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201...203
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10197

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10197
Availability details for
Quantity
  • Availability
  • Add to Cart