ENA024296235 Enamine

N-[4-(chloromethyl)-1,3-thiazol-2-yl]-N-(3-methylphenyl)acetamide

  • MDL number MFCD06341990
  • Cas Number : 756859-01-5
  • Molecular Weight : 280.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-91
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10199
Availability details for
Quantity
  • Availability
  • Add to Cart