ENA024296237 Enamine

2-[(2-chlorobenzoyl)amino]-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid

  • MDL number MFCD01238951
  • Cas Number : 326611-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10201

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10201
Availability details for
Quantity
  • Availability
  • Add to Cart