ENA024296255 Enamine

2-chloro-N-{[(2,4,6-trimethylphenyl)carbamoyl]methyl}acetamide

  • MDL number MFCD06342746
  • Cas Number : 757220-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10219
Availability details for
Quantity
  • Availability
  • Add to Cart