ENA024296259 Enamine

2-chloro-5-{[4-(trifluoromethoxy)phenyl]sulfamoyl}benzoic acid

  • MDL number MFCD06334884
  • Cas Number : 745798-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10223
Availability details for
Quantity
  • Availability
  • Add to Cart