ENA024296279 Enamine

2-chloro-N-[3,5-dimethyl-1-(4-methylphenyl)-1H-pyrazol-4-yl]acetamide

  • MDL number MFCD06342757
  • Cas Number : 956743-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10243
Availability details for
Quantity
  • Availability
  • Add to Cart