ENA024296294 Enamine

2-[(2-ethylbutanoyl)amino]-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid

  • MDL number MFCD06342760
  • Cas Number : 757220-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10258

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10258
Availability details for
Quantity
  • Availability
  • Add to Cart