ENA024296304 Enamine

2-({[(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)carbamoyl]methyl}sulfanyl)acetic acid

  • MDL number MFCD02680096
  • Cas Number : 697741-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-165
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10268
Availability details for
Quantity
  • Availability
  • Add to Cart