ENA024296305 Enamine

4-amino-N-methyl-N-(propan-2-yl)benzene-1-sulfonamide

  • MDL number MFCD06342776
  • Cas Number : 757221-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-99
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10269

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10269
Availability details for
Quantity
  • Availability
  • Add to Cart