ENA024296317 Enamine

2-methyl-2-(methylamino)propanenitrile

  • MDL number MFCD03208517
  • Cas Number : 2273-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10281

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P264,P270,P280,P301+P310,P303+P361+P353,P330,P370+P378,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-10281
pictograms GHS02,GHS06
hstatements H226,H301
Availability details for
Quantity
  • Availability
  • Add to Cart