ENA024296329 Enamine

4-({[4-chloro-3-(trifluoromethyl)phenyl]sulfonyl}amino)butanoic acid

  • MDL number MFCD03786818
  • Cas Number : 612043-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10293

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10293
Availability details for
Quantity
  • Availability
  • Add to Cart